Magnetic Seals w/Septa, 20 mm, preassembled, Restek
Supplier: Restek
22441 22832 22833 22836 22442 22443 22444 22831 22834 22835
10058-778PK 114.51 USD
10058-778 10058-798 10058-800 10058-802 10854-116 10854-118 10854-120 10854-154 10854-156 10854-158
Magnetic Seals w/Septa, 20 mm, preassembled, Restek
Closures Chromatography Vial Closures
Magnetic seals with variable mouth sizes are compatible with a range of vials with 20-millimeter orifices.
  • Aluminum/steel or steel fabrication
  • Compatible with crimp-top vials
  • 8 mm opening
BiMetal magnetical seals are easier to crimp and decap than standard 20 mm magnetic seals.
Ordering information: 5 mm hole is compatible with the following systems: Carlo Erba HS500/HS800, CTC 500, Fisons HS500/HS800, and Bruker/Varian/Chrompack 9020/25. 8 mm hole is compatible with the Combi Pal.