Nut Driver, ¹/₄", Restek
Supplier: Restek
21076
10058-596EA 43.57 USD
10058-596
Nut Driver, ¹/₄", Restek
Nutdrivers
Ideal for changing Agilent 5890/6850/6890 FID jets.
  • Securely grips jet for easy removal or installation