Centrex™ Centrifuge Filters, Maine Manufacturing
Supplier: GVS FILTER TECHNOLOGY
10467009
10057-608PK 1596.41 USD
10057-608
Centrex™ Centrifuge Filters, Maine Manufacturing
Filters Centrifugal Filters
Centrifugal filter units with membrane filter for rapid and simple preparation of a large number of samples.
  • More than six samples can be processed at once
  • Ideal for automated systems and high-speed batch filtration with robots
  • Considerably reduced contamination risk when working with radioactive biologically hazardous material
  • Cross contamination avoided
  • Receiver tubes 1.5 or 5 ml
Cellulose acetate membrane (0.45 μm) for the rapid elution of agarose gels. Nylon and cellulose acetate membranes for the removal of particles and microorganisms from HPLC samples, sample preparation for quality control, cellulose acetate and nitrocellulose membrane for rapid clearing and filtration of aqueous solutions.