GC Column Selection by U.S EPA Drinking Water Test Method, Thermo Scientific
Supplier: Avantor
10057-020EA 1068.35 USD
10057-020 10057-022
GC Column Selection by U.S EPA Drinking Water Test Method, Thermo Scientific
Chromatography Columns
These columns are designed to perform specific EPA methodologies. These low-bleed and temperature-stable columns have been designed, manufactured and tested to provide consistent, reliable results from run-to-run and batch-to-batch.
  • Low bleed, even at elevated temperatures
  • Outstanding robustness for difficult separations
  • Reliable and reproducible performance
Applications: Volatile organic compounds (VOCs), pesticides, brominated flame retardants, explosives.