TouchNTuff® 73-300 Neoprene Gloves, Non-Sterile, Ansell
Supplier: Ansell Healthcare
73300055 73300060 73300065 73300070 73300075 73300080 73300090 73300085
10054-568CS 156.32 USD
10054-568 10054-570 10054-572 10054-574 10054-576 10054-578 10054-582 76415-412
TouchNTuff® 73-300 Neoprene Gloves, Non-Sterile, Ansell
Gloves
The TouchNTuff® 73-300 neoprene gloves deliver superior sensitivity and comfort in nonsterile, clean environments.
  • Hand-specific
  • Great splash resistance against a broad range of chemicals
  • Ergonomically designed to enhance fit and comfort
  • Reduced hand fatigue and stress
These sensitive neoprene gloves are a new feel in double-donning and dry applications.
TouchNTuff® single-use gloves provide superior tactility and high resilience in wet and dry industrial, lab, and controlled environments. Gloves are packed in a controlled environment, in a cleanroom-compatible packaging material, which is printed with IPA-resistant ink.
Customers need to make their own determination based on cleanliness standards. Ansell expects this glove should be suitable for ISO Class 5 or higher cleanroom environments.
The exceptionally thin polychloroprene formulation reduces the risk of Type I or IV allergies to the wearer caused by natural rubber latex or other synthetic gloves. The sensitive neoprene gloves offer users splash resistance against a broad range of chemicals. Ansell’s SureFit Technology™ eliminates 'cuff roll down' by maintaining gloves securely around the arm, providing wearer with more protection during routine lab work and manufacturing by ensuring the efficacy of the gloves as a barrier protection. Gloves are ergonomically designed and hand-specific to enhance fit and comfort, and to reduce hand fatigue and stress.
Certifications: Audit Standards Manufacturing Facility Registered to ISO 9001, ISO 13485 and ISO 14001. Quality Control Compliant with ASTM D 3577, Type II, Cat. III glove.