Heating Shaking Dry Bath Incubator, Thermo Scientific

Supplier: Thermo Fisher Scientific
88880027
10054-216EA 2433.31 USD
10054-216
Heating Shaking Dry Bath Incubator, Thermo Scientific
Shakers and Mixers Shaking Incubators
Increase lab versatility with the Thermo Scientific™ Digital Heating Shaking Drybath, which is easy to use and can be used as a drybath, shaker or both. Precisely control both temperature and shaking speed digitally, for accurate repeatable results.

  • PID control ensures consistent shaking results and precise temperature control to 0.1 degree
  • Speed range from 150 to 1500 rpm (block dependent)
  • Program Storage 10 programs, up to 10 steps per program
  • Interval shaking function allows shaking and non-shaking in same program
  • Two types of timing modes: Start only after temperature reaches desired set point; or Start immediately after the timer is set
  • Low-profile design
  • Easily changed aluminum blocks in multiple sizes allow for maximum flexibility in vessel choice
  • Over temperature cut off safety protection

Precise accuracy is ensured (0.1 °C) by its PID controller and timer. Usable with 96-well, 0.5 ml, 1.5 ml, and 50 ml tubes, blocks purchased separately. Wide temperature range, Ambient +5 to –100 ℃ covers a broad range of applications including thawing, incubation, enzyme reactions, blood urea nitrogen determinations, and immunoassays.

Orbital diameter: (mm/Inch) 2/0.08
Uniformity(±°C/°F) 0.5/0.9 at 80 °C
Stability(±°C/°F) 0.15/0.27 at 80 °C

Certifications: CE

Ordering information: Must purchase a block with unit.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR