RT Touch Series Magnetic Stirrers, Thermo Scientific
Supplier: Thermo Fisher Scientific
88880015
10054-188EA 627.67 USD
10054-188 10054-192
RT Touch Series Magnetic Stirrers, Thermo Scientific
Stirrers Magnetic Stirrers
The RT touch digital stirrers feature a chemical-resistant polypropylene top and body, a strong magnetic coupling to ensure consistent stirring during experimental procedures, a timer, and a brushless DC Motor that will not generate heat.
  • Precise digital control capable of slow speed stirring
  • Timer runs continuous or countdown cycles, low-profile, lightweight design with small footprint
  • Strong magnetic coupling ensures consistent stirring during experimental procedures
  • Brushless dc motor will not generate heat and 'cook' samples
  • Polypropylene top and body provide excellent chemical resistance
  • Two plate sizes available: 170 mm, or 220 mm and detachable plug
  • Two non-slip silicone plate covers included (1 black, 1 white); Gradual start-up stirring to prevent splashing
  • Threaded hole for support rod; Low power consumption
Experience precise control with the RT Touch series digital magnetic stirrers. Precise digital speed control ranges from 30 to 2000 rpm. Two plate sizes are available (170 mm or 220 mm) to choose from, each with a low-profile, lightweight design and an extremely small footprint. All units include two non-slip silicone plate covers for added protection from spills.
Certifications: CE marked.
Ordering information: 2-Year warranty.