Liquid Sampling Kit, Dynalon
Supplier: Dynalon
107035-3000
10053-758EA 496.41 USD
10053-758
Liquid Sampling Kit, Dynalon
Samplers Fluid Samplers
These versatile modular design plastic liquid sampling kits will meet all of your needs
Kit components are manufactured of high density polyethylene (HDPE) and polypropylene (PP). Their threaded connections allow quick and easy use. The Liquid Sampling Kit includes a sampler component with 100ml and 500ml polypropylene bottle plus a 1.5', 3' and 6' HDPE handle.
Whether sampling from drums, tanks, or rail cars, taking water samples or dipping chemicals from a bath we have a kit configuration to meet your liquid sampling application requirements.
Weight: 5.1 lbs.
Certifications: FDA Compliant.
Ordering information: Liquid Sampling Kit components are interchangeable. Component are also sold as separate items (See 10053-764, -766, -768, -750 and -752). Kits and their components are available as one each.