Stirring Kit, Dynalon
Supplier: Dynalon
107035-2000
10053-756EA 341.23 USD
10053-756
Stirring Kit, Dynalon
Stirrers Stirring Rods
These versatile modular design plastic stirring kits will meet all of your needs
Kit components are manufactured of high density polyethylene (HDPE). Their threaded connections allow quick and easy use. The Stirring Kit includes a stirring and perforated mixing paddle plus a 1.5', 3' and 6' handle.
Whether stirring in product vats, mixing in a tank or stirring chemicals from a bath we have a kit configuration to meet your stirring and mixing application requirements.
Certifications: FDA Compliant.
Ordering information: Stirring Kit components are interchangeable. Component are also sold as separate items (See 10053-764, -766, -768, -776 and -778). Kits and their components are available as one each.