ASTM-Like Non-Mercury Thermometers, Thermco
Supplier: Thermco
ACC001CBLS ACC001FBLS ACC009CS ACC038CBLS ACC039CBLS ACC040CBLS ACC042CBLS ACC098CBLS ACC099CBLS ACC014CBLS ACC014FBLS ACC015CBLS ACC015FBLS ACC016FBLS ACC017CBLS ACC017FBLS ACC019CBLS ACC019FBLS ACC020CBLS ACC020FBLS ACC099FBLS ACC113CBLS ACC113FBLS ACC021CBLS ACC033CBLS ACC033FBLS ACC034CBLS ACC034FBLS ACC036CBLS ACC037CBLS ACC049CBLS ACC052CBLS ACC054CBLS ACC054FBLS ACC057CBLS ACC057FBLS ACC061CBLS ACC061FBLS ACC083CBLS ACC083FBLS ACC084CBLS ACC084FBLS ACC085CBLS ACC085FBLS ACC088CBLS ACC088FBLS ACC089CBLS ACC090CBLS ACC097CBLS ACC097FBLS
10053-014EA 183.21 USD
10053-014 10053-016 10053-018 10053-020 10053-022 10053-024 10053-026 10053-028 10053-030 10053-032 10053-034 10053-244 10053-246 10053-248 10053-250 10053-252 10053-254 10053-256 10053-258 10053-260 10053-262 10053-264 10053-266 10053-460 10053-462 10053-464 10053-612 10053-614 10053-616 10053-618 10053-620 10053-622 10053-624 10053-626 10053-628 10053-630 10053-632 10053-634 10053-636 10053-638 10053-640 10053-642 10053-648 10053-650 10053-652 10053-654 10053-656 10053-658 10053-664 10053-666 10053-668 10053-670 10053-672 10053-674
ASTM-Like Non-Mercury Thermometers, Thermco
Thermometers Analog Thermometers
ASTM-like non-mercury blue spirit "precision" thermometers with the same specifications as the mercury ASTM thermometers
Each is designated for an ASTM method, serialized, filled with a blue non-mercury liquid with a white background for easy reading of temperature values.
Certifications: ASTM approved/ISO17025/Statement of Traceability.
Ordering information: Hard plastic square tube included.