ASTM Approved Non-Mercury Thermometers, Thermco
Supplier: Thermco
ACC005CBLS ACC005FBLS ACC012CBLS ACC012FBLS ACC018CBLS ACC018FBLS ACC022CBLS ACC022FBLS ACC117CBLS ACC120CBLS ACC130CBLS ACC130FBLS ACC056CBLS ACC056FBLS ACC058CBLS ACC058FBLS ACC059FBLS ACC091CBLS ACC116CBLS
10052-990EA 157.8 USD
10052-990 10052-992 10052-994 10052-996 10052-998 10053-000 10053-002 10053-004 10053-006 10053-008 10053-010 10053-012 10053-228 10053-230 10053-232 10053-234 10053-236 10053-238 10053-240 10053-242
ASTM Approved Non-Mercury Thermometers, Thermco
Thermometers Analog Thermometers
ASTM approved non-mercury blue spirit "precision" thermometers with the same specifications as the mercury ASTM thermometers
Each is designated for an ASTM method, serialized, filled with a blue non-mercury liquid with a white background for easy reading of temperature values.
Certifications: ASTM approved/ISO17025/Statement of Traceability.
Ordering information: Hard plastic square tube included.