DM1000 LED Upright Microscope Package, Leica Microsystems
Supplier: Avantor
11591301 11505299
10037-580EA 3985.26 USD
10037-580 10052-384 76382-984 76382-986 76414-908
DM1000 LED Upright Microscope Package, Leica Microsystems
Microscopes
The DM1000 LED microscope is designed for applications in the biological, medical and clinical laboratories; from routine tasks to research.
  • Upright microscope with complete adjustability
  • High-performance optics
  • Ergonomic design
  • Long lasting LED illumination
  • HC Plan 10×/20 BR eyepiece
The DM1000 LED microscope lab package is a robust upright microscope with LED illumination designed for years of use without the need to replace bulbs.
The DM1000 LED makes routine work as comfortable and effortless as possible. It is completely adjustable to each individual user including height-adjustable focus knobs, adjustable tubes, symmetrical operation, and fast changeover from right- to left-handed operation. The DM1000 LED features Hi Plan objectives including 4×, 10×, 20×, 40× and 100×. Additional N Plan objective can be added for additional imaging capabilities. When properly configured, the DM1000 LED can be combined with a selection of cameras from the Leica camera portfolio. These include the 5 MP MC170, the 3 MP DMC2900 and the 1.3 MP DFC3000 G.
Additional accessories are available to fit most imaging applications.
Certifications: C-UL (US) and CE.
Ordering information: When ordering the DM1000 LED with a camera please ensure that you select at least one of the three camera choices and the optional imaging module. The DM1000 LED imaging solution will come with a 1-year limited warranty.