Chromium Standards, CRM for AA and ICP, Assurance Grade, SPEX CertiPrep®
Supplier: SPEX CERTIPREP GROUP LLC MS
SPEC-CR6
100515-431EA 80.77 USD
100515-431
Chromium Standards, CRM for AA and ICP, Assurance Grade, SPEX CertiPrep®
Standards Cationic Standards Chromium Standards
  • Traceable to NIST
  • Uncertainty of standards is reported on the Certificate of Analysis
  • Manufactured by experienced chemists to ensure highest quality standards
Caution: 1000 µg/mL = 1000 ppm