Mouse IgG2b Monoclonal Isotype Control [clone: MPC-11] (violetFluor® 450)

Supplier: Tonbo Biosciences
75-4732-U100
10051-856EA 141.9 USD
10051-856
Mouse IgG2b Monoclonal Isotype Control [clone: MPC-11] (violetFluor® 450)
Antibodies
The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.

Type: Control
Antigen: MPC11
Clonality: Monoclonal
Clone: MPC-11
Conjugation: violetFluor® 450
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR