4-Hydroxy-2-quinolinecarboxylic acid

Supplier: R&D Systems

Synonyms: 4-Hydroxyquinoline-2-carboxylic Acid, Transtorine, Empirical Formula, Kynurenic acid, Quinurenic acid, KYNA, Kynuronic acid, Kinurenic acid

0223/1G
100506-418EA 163.42 USD
100506-418
4-Hydroxy-2-quinolinecarboxylic acid
4-Hydroxy-2-quinolinecarboxylic acid
Formula: C₁₀H₇NO₃
Melting Pt: 275 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149476
CAS Number: 492-27-3
EINECS: 207-751-5
Merck Index: 13,05342

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR