Capsaicin
Supplier: R&D Systems
Synonyms:
8-Methyl-N-vanillyl-6-none-amid
0462/100
100506-070EA
267.54
USD
100506-070
Capsaicin
Capsaicin
Formula:
C₁₈H₂₇NO₃ MW: 305.42 g/mol Boiling Pt: 210…220 °C (0.01 mmHg) Melting Pt: 66.1 °C |
MDL Number:
MFCD00017259 CAS Number: 404-86-4 EINECS: 206-969-8 UN: 2811 ADR: 6.1,III Merck Index: 12,01811 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...