Dexamethasone

Supplier: R&D Systems

1126/100
100505-728EA 163.42 USD
100505-728
Dexamethasone
Dexamethasone
Formula: C₂₂H₂₉FO₅
Melting Pt: 268…271 °C
MDL Number: MFCD00064136
CAS Number: 50-02-2
EINECS: 200-003-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR