4-Aminopyridine

Supplier: R&D Systems

0940/100
100505-204EA 156.18 USD
100505-204
4-Aminopyridine
4-Aminopyridine
Formula: C₅H₆N₂
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 157…160 °C
Density: 1.25 g/cm³ (20 °C)
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00006439
CAS Number: 504-24-5
EINECS: 207-987-9
UN: 2671
ADR: 6.1,II
Merck Index: 13,03964

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR