Glycine

Supplier: R&D Systems

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

0219/1G
100505-088EA 117.14 USD
100505-088
Glycine
Glycine
Formula: NH₂CH₂COOH
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 83 °C
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR