Nigrosine, certified (C.I. 50420)

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Acid Black 2

19010
100505-026EA 62.37 USD
100505-026
Nigrosine, certified (C.I. 50420)
Nigrosine

For negative staining of bacteria. With Basic Fuchsin, staining bacteria spores. Dorner, Le Lait, 6, 8 (1926). Biochem. Biophys. Acta 69,159 (1963).


Solubility : 10.0% Water; 0.0% Alcohol; 1.0% Cellosolve; 11.0% Glycol; 0.0% Xylene

Melting Pt: 275 °C (527 °F)
Storage Temperature: Ambient
MDL Number: MFCD00044681
CAS Number: 8005-03-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc4b175e0f3858b65e354c84d7c4e792cb94c425&vid=fc4b175e0f3858b65e354c84d7c4e792cb94c425&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR