Ampoule Breaker, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60600
100504-972EA 36.89 USD
100504-972
Ampoule Breaker, Electron Microscopy Sciences
Ampule Openers
This reausable ampoule breaker is made from soft silicone.
  • Accommodates 5, 7 and 10 mL size ampoules
  • Reduces injury and spillage
This provides a simple way to break pre-scored glass ampoules safely.