Sodium methoxide, anhydrous

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Sodium methylate, Sodium methanolate

21200
100504-966EA 20.97 USD
100504-966
Sodium methoxide, anhydrous
Sodium methoxide

Sodium methoxide can used in the removal of resin from sections.

Formula: CH₃NaO
Density: 0.97 g/cm³ (8.095 lbs/gal.)
Flash Pt: 12 °C (54 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012179
CAS Number: 124-41-4
UN: 1431
ADR: 4.2,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bffaf4ba73f73a65dc485fb954252182a77955c8&vid=bffaf4ba73f73a65dc485fb954252182a77955c8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR