2-Methylbutane, Reagent Grade

Supplier: Electron Microscopy Sciences
Danger

Synonyms: iso-Pentane

18550
100504-954EA 94.47 USD
100504-954
2-Methylbutane, Reagent Grade
2-Methylbutane

(Isopropentane) CH2H5CH(CH3)2 F.W. 72.15 CAS 78-78-4 b.p. 27-31°C Specific Gravity 20°C: 0.62

Formula: CH₃CH₂CH(CH₃)₂
MW: 72.15 g/mol
Boiling Pt: 27.95 °C (82 °F)
Melting Pt: –158.5 °C (–253 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009338
CAS Number: 78-78-4
UN: 1265
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98d4a4b81f477c99371c19cfa1ac9f6e293b186b&vid=98d4a4b81f477c99371c19cfa1ac9f6e293b186b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Specific Gravity at 20 °C 0.62

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR