Methacrylic acid glacial ≥98% stabilised

Supplier: Electron Microscopy Sciences
Danger

Synonyms: 2-Methylpropenoic acid

Stabilization: 0.25% Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)

18500
100504-952EA 68.54 USD
100504-952
Methacrylic acid glacial ≥98% stabilised
Methacrylic acid

(2-Methylpropenoic Acid) CH2:C(CH3)COOH F.W. 86.09 Assay: 98% Stab. with 0.25% Hydroquinone monomethyl ether.

Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 163
Melting Pt: 15
Density: 1.0153 g/cm³ (8.473 lbs/gal.)
Flash Pt: 68 °C (154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002651
CAS Number: 79-41-4
UN: 2531
ADR: 8 II
Merck Index: 13,05967
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d95a7e7baebcd24667a09dc6a945f5f7d8d328b&vid=c8cad964a1c0ddd1ff34ffe4b5944fb2a3d15f4a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay >98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR