Toluidine blue O, certified (C.I. 52040)

Supplier: Electron Microscopy Sciences
Warning

Synonyms: TBO, 3-Amino-7-(dimethylamino)-2-methyl-5-phenothiazinium chloride, 3-Amino-7-(dimethylamino)-2-methylphenothiazin-5-ium chloride, Tolonium chloride, Basic Blue 17, Blutene chloride

22050
100504-874EA 72.88 USD
100504-874
Toluidine blue O, certified (C.I. 52040)
Toluidine blue O

Toluidine blue 0 can be used as a general stain for epoxy thick sections (Barajas et al.(1981)).


Identification of renal neuroeffector junctions is by electron microscopy of reembedded light microscopic autoradiograms of semithin sections (J. Ultrastruct. Res. 77: 379.Campbell, R.D., And Hermans, C.O. (1972)).
A rapid method for resectioning 0.5 4.0 micron epoxy sections for electron microscopy (Stain Technol . 47:115.).

Formula: C₁₅H₁₆ClN₃S
Boiling Pt: 100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011934
CAS Number: 92-31-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c7665296e38e6a43633d044660505caf7ca911c&vid=4c7665296e38e6a43633d044660505caf7ca911c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Dye content approx. 85% min


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR