Calcium chloride dihydrate ACS

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Calcium Chloride Dihydrate, Calcium dichloride

12340
100504-842EA 39.3 USD
100504-842
Calcium chloride dihydrate ACS
Calcium chloride dihydrate

CaCl2·2H2O F.W. 147.02 CAS 10035-04-8

Formula: CaCl₂·2H₂O
MW: 147.01 g/mol
MDL Number: MFCD00149613
CAS Number: 10035-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR