Potassium hexacyanoferrate(III) solution 5% (Johnson's Method for Iron)

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide

26603-01
100504-670EA 49.28 USD
100504-670
Potassium hexacyanoferrate(III) solution 5% (Johnson's Method for Iron)
Potassium hexacyanoferrate (III)

Johnson's Method for Iron - (Microincineration). Mineral and Pigment stain.


Stain results - Iron Oxide: Blue.


References:
Fenton R.H., Johnson, F.B., and Zimmerman, L.E., J. Histo-chem. Cytochem., 12:153-155, 1964.
Luna, L.G., (ed.), Manual of Histologic Staining Methods of the AFIP, 3rd edition, McGraw-Hill, NY, p.181, c 1968.

Formula: K₃Fe(CN)₆
MW: 329.25 g/mol
MDL Number: MFCD00011392
CAS Number: 13746-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR