Direct red 80 0.1% in saturated picric acid

Supplier: Electron Microscopy Sciences
Warning

Synonyms: CI 35780, Sirius Red F3B, Sirius Red, Picro-Sirius Red

26357-02
100504-596EA 26.06 USD
100504-596
Direct red 80 0.1% in saturated picric acid
Direct Red 80

Picrosirius Red Staining of Cardiac Muscle Following Phosphomolybdic Acid Treatment.
For connective tissue stains – Van Gieson stains and variants. Septa is stained red.


This method eliminates cytoplasmic staining, revealing thin septa and collegen fibers clearly.
Fixation: Bouins' Fixative
Reagents
26357-01 Phosphomolybdic Acid, 0.2% Aqueous
26357-02 Sirius Red (Direct Red 80), 0.1% in Saturated Picric Acid
26357-03 Hydrochloric Acid, 0.01N


References: Dolber, Paul C. and Madison S. Spach. Picrosirus Red Staining of Cardiac Muscle Following Phosphomolybdic Acid Treatment, Stain Technology, Vol. 62 No 1 (1987) 23 – 26.


Certifications: Certificates of analysis are available from Electron Microscopy Sciences. Please contact us or visit our website to search by lot or product number.

Formula: C₄₅H₂₆N₁₀Na₆O₂₁S₆
MW: 1373.051 g/mol
MDL Number: MFCD00054389
CAS Number: 2610-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR