Crystal violet staining solution, for amyloid in histology and pathology

Supplier: Electron Microscopy Sciences

26301-01
100504-578EA 106.93 USD
100504-578
Crystal violet staining solution, for amyloid in histology and pathology
Crystal Violet

Stain results:
Amyloid: Violet-Red
Tissue: Blue


To combine with Acetic Acid, 1% Aqueous (26301-02)

Formula: C₂₅H₃₀ClN₃
MDL Number: MFCD00011750
CAS Number: 548-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR