Giemsa's stain stock solution for Helicobactor Pylori and Mast Cells

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

26110-02
100504-522EA 31.49 USD
100504-522
Giemsa's stain stock solution for Helicobactor Pylori and Mast Cells
Giemsa's stain

For bacterial, fungal stains.


References:
Helicobactor pylori in peptic ulcer disease, NIH Consensus Statement Online, January 1994, 12(1): 1:23
Luna 1AFIP 3rd ed. McGraw Hill, 236
Fail, Rena, AS, HT (ASCP) Medical University of South Carolina, Histologic. Vol. XXXIV, No 2, Nov. 2001, p. 32.

MDL Number: MFCD00081642
CAS Number: 51811-82-6
UN: 1230
ADR: 3 (6.1),II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR