Methanol for Helicobactor Pylori and Mast Cells

Supplier: Electron Microscopy Sciences
Danger

26110-01
100504-520EA 27.51 USD
100504-520
Methanol for Helicobactor Pylori and Mast Cells
Methanol

used in Giemsa for Helicobactor Pylori and Mast Cells.

Formula: CH₃OH
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3 (6.1),II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR