Cellosolve for bacteriology (Brown-Hopps method)

Supplier: Electron Microscopy Sciences
Warning

26106-05
100504-504EA 54.8 USD
100504-504
Cellosolve for bacteriology (Brown-Hopps method)
2-Ethoxyethanol

Cellosolve, Brown-Hopps Method reagent for Gram Positive and Gram Negative Bacteria.


Stain results:
Gram Negative Bacteria: Red
Gram Positive Bacteria: Blue
Background: Yellow

Formula: C₂H₅OCH₂CH₂OH
Boiling Pt: 135 °C (275 °F)
Melting Pt: –100 °C (–148 °F)
Density: 0.93 g/cm³ (7.761 lbs/gal.)
Flash Pt: 40 °C (104 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002869
CAS Number: 110-80-5
EINECS: 203-804-1
UN: 1171
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=461a4daf8709ebd6b4516d73b4b32eee33cd1bca&vid=625adefb8d1e2126363931c57a51d3e1df419926&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR