Basic fuchsin 0.5% in aqueous solution staining solution, for bacteriology (Brown-Hopps method)

Supplier: Electron Microscopy Sciences
Danger

Synonyms: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline

26106-03
100504-500EA 64.43 USD
100504-500
Basic fuchsin 0.5% in aqueous solution staining solution, for bacteriology (Brown-Hopps method)
Basic Fuchsin

Brown-Hopps Method reagent for Gram Positive and Gram Negative Bacteria.


Stain results:
Gram Negative Bacteria: Red
Gram Positive Bacteria: Blue
Background: Yellow

Formula: C₂₀H₂₀ClN₃
MW: 337.85 g/mol
MDL Number: MFCD00012569
CAS Number: 632-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR