Crystal violet 1% in aqueous solution stain for bacteriology (Brown-Hopps method)

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet

26106-01
100504-496EA 41.39 USD
100504-496
Crystal violet 1% in aqueous solution stain for bacteriology (Brown-Hopps method)
Crystal Violet

Brown-Hopps Method reagent for Gram Positive and Gram Negative Bacteria.


Stain results:
Gram Negative Bacteria: Red
Gram Positive Bacteria: Blue
Background: Yellow

Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
MDL Number: MFCD00011750
CAS Number: 548-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR