Tritolyl phosphate (mixture of isomers)

Supplier: Electron Microscopy Sciences
Warning

22150
100504-330EA 39.77 USD
100504-330
Tritolyl phosphate (mixture of isomers)
Tritolyl phosphate (mixture of isomeres)

A mixture of isomeric tritolyl phosphates, usually excluding the very toxic group ortho-isomer.


Used to prepare benzoyl peroxide paste, which acts as a catalyst for water miscible resins.

Formula: C₂₁H₂₁O₄P
Density: 1.143 g/cm³ (9.538 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD02102116
CAS Number: 1330-78-5
UN: 2574
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6039494cf0bab71b3753d169290a40e34d441d15&vid=6039494cf0bab71b3753d169290a40e34d441d15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Specific gravity (H₂O=1) 1.16


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR