Tetramethylsilane ≥99.9%

Supplier: Electron Microscopy Sciences
Danger

Synonyms: TMS

21760
100504-314EA 319.32 USD
100504-314
Tetramethylsilane ≥99.9%
Tetramethylsilane

An organo-silicon compound tetramethylsilane (TMS) is used as an internal standard for nuclear magnetic resonance (NMR).


TMS is an ideal solvent for tissue drying, which preserves excellent tissue surface (Dey, Sudip. Et al. (1989)). A new rapid method of air-drying for scanning electron microscopy is possible with tetramethylsilane (J. of Microscopy, Vol 156, pp. 259-261).

Formula: C₄H₁₂Si
MW: 88.22 g/mol
Boiling Pt: 26.6 °C
Density: 0.648 g/cm³ (5.408 lbs/gal.)
Flash Pt: –54 °C (–65 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008274
CAS Number: 75-76-3
UN: 2749
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05a9323978afbdeab3cbeba3a8660938f7de4f16&vid=05a9323978afbdeab3cbeba3a8660938f7de4f16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR