Succinic acid ≥99.0%, crystals ACS

Supplier: Electron Microscopy Sciences
Danger

Synonyms: 1,4-Butanedioic acid

21550
100504-302EA 51.71 USD
100504-302
Succinic acid ≥99.0%, crystals ACS
Succinic acid

(CH3COOH)2 F.W. 118.09x m.p. 185-191°C Assay 99.0%

Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Boiling Pt: 235 °C (455 °F)
Melting Pt: 184 °C (363 °F)
Density: 1.564 g/cm³ (13.052 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b9b5e7c10264cd49274179d5192aff06ada8d29&vid=7b9b5e7c10264cd49274179d5192aff06ada8d29&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0%
Chloride 0.001%
Heavy metals 5 ppm
Insoluble matters 0.01%
Iron 5 ppm
Nitrogen compounds 0.001%
Phosphate 0.001%
Sulfate 0.003%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR