di-Sodium L(+)-tartrate dihydrate 99.5-101.0% ACS

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Sodium L-(+)-tartrate dihydrate, L(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Sodium tartrate dibasic dihydrate, Disodium L-(+)-Tartrate Dihydrate, (R,R)-(+)-Disodium tartrate dihydrate, L(+)-Tartaric acid, disodium salt, dihydrate, Sodium L-tartrate dibasic dihydrate, L(+)-Sodium tartrate dibasic dihydrate, Sodium Tartrate, Dihydrate, (+)-Disodium L-tartrate dihydrate, (+)-Disodium-L-tartrat dihydrate

21350
100504-296EA 25.68 USD
100504-296
di-Sodium L(+)-tartrate dihydrate 99.5-101.0% ACS
di-Sodium L(+)-tartrate dihydrate

Na2C4H4O6·2H2O F.W. 230.08 Assay 99.5%-101.0% pH of a 5% solution 25°C-7.0-9.0 Loss on Drying 150°C-15.61-15.71%

Formula: NaOOC(CHOH)₂COONa·2H₂O
MW: 230.08 g/mol
Melting Pt: 150 °C (302 °F)
Density: 1.82 g/cm³ (15.188 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00150035
CAS Number: 6106-24-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bc26a812e504254c397ae23b354b3d5dbba4804&vid=9bc26a812e504254c397ae23b354b3d5dbba4804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Ammonium 0.003%
Assay 99.5%-101.0%
Calcium 0.01%
Chloride 0.0005%
Heavy metals 0.0005%
Insoluble matters 0.005%
Iron 0.001%
Loss on drying (150 °C) 15.61-15.71%
pH of 5% solution (25 °C) 7.0-9.0
Phosphate 0.0005%
Sulfate 0.005%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR