Sodium sulfite ≥98.0% ACS

Supplier: Electron Microscopy Sciences
Warning

21300
100504-294EA 106.74 USD
100504-294
Sodium sulfite ≥98.0% ACS
Sodium sulphite

Assay 98.0%

Formula: Na₂SO₃
MW: 126.04 g/mol
Density: 2.63 g/cm³ (21.947 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=922353b9006f619320aa4c1df3a152577e7e092b&vid=922353b9006f619320aa4c1df3a152577e7e092b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Insoluble matters 0.005%
Free acid To pass test
Titratable free base 0.03 meq/g
Chloride 0.02%
Arsenic 0.0001%
Heavy metals 0.001%
Iron 0.001%
Assay 98.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR