Sodium ethoxide

Supplier: Electron Microscopy Sciences
Danger

21150
100504-280EA 25.19 USD
100504-280
Sodium ethoxide
Sodium ethoxide

Used as an agent for flattening thick sections (0.5 to 2.0 micron) of epoxy embedded material (Felix Sierra, Christa Hegele-Hartung and Henning M. Beier (1990)).


An improved technique for flattening semithin sections of epoxy-embedded tissues used for high resolution light microscopy (J. of Microscopy Vol 159, Pt 1, (July 1990), pp. 95-98).

Formula: C₂H₅NaO
Density: 0.88 g/cm³ (7.344 lbs/gal.)
Flash Pt: 30 °C (86 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012417
CAS Number: 141-52-6
UN: 3206
ADR: 4.2,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68a3915dfc55311edde61bbc9ed6bcdaa1580a2e&vid=68a3915dfc55311edde61bbc9ed6bcdaa1580a2e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR