di-Sodium tetraborate decahydrate 99.5-105.0% ACS

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate

21130
100504-274EA 71.81 USD
100504-274
di-Sodium tetraborate decahydrate 99.5-105.0% ACS
di-Sodium tetraborate decahydrate

(Sodium Tetraborate, Decahydrate) Na2B4O7.10H2O FW. 381.37 CAS 1303-96-4 A.C.S.

Formula: Na₂B₄O₇·10H₂O
MDL Number: MFCD00149193
CAS Number: 1303-96-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49c9a6ae89a98c5766d3dde9280bceef949428dc&vid=49c9a6ae89a98c5766d3dde9280bceef949428dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5 - 105%
Calcium 0.005%
Chloride 0.001%
Insoluble Matters 0.005%
Iron 5 ppm
Other Heavy Metals (as Pb) 0.001%
pH of 0.01 M Solution at 25 °C 9.15 - 9.20
Phosphate 0.001%
Sulfate 0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR