Sodium acetate trihydrate 99.0-100.5% ACS

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Acetic acid sodium salt trihydrate

21120
100504-272EA 96.73 USD
100504-272
Sodium acetate trihydrate 99.0-100.5% ACS
Sodium acetate trihydrate

CH3COONa·3H2O F.W. 136.08 A.C.S.

Formula: H₃CCOONa·3H₂O
Melting Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e84caa08c81543a06fc8acc31f6ecc6b4f084682&vid=e84caa08c81543a06fc8acc31f6ecc6b4f084682&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0-100.5%
Ca, Mg and R₂O₃ ppt 0.01%
Chloride 0.001%
Insoluble matters 0.005%
Iron, heavy metal, phosphate 5 ppm/ea
pH of 5% solution (25 °C) 7.5-9.2
Potassium 0.005%
Sulfate 0.002%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR