Safranine O, certified (C.I. 50240)

Supplier: Electron Microscopy Sciences
Warning

20800
100504-260EA 73.93 USD
100504-260
Safranine O, certified (C.I. 50240)
Safranine O

(Safranin T, Y or A; Basis Red 2; Cotton Red; Gossypimine) C20H19N4Cl F.W. 350.85 Solubility: 4.5% Water; 3.5% Alcohol; 5.0% Cellosolve; 3.5% Glycol; 0.0% XylenePrussian Blue method for hemosiderin. Weigert's iron hematoxylin with methachromic dyes which stain nuclei and granules. A alkaline phosphate stain. Flemming's method for staining chromatin and nuclear elements. Certified by the Biological Stain Commission All Dyes/Stains are for research use only. Not meant for drug, food household use.


Solubility : Water; 3.5% Alcohol; 5.0% Cellosolve; 3.5% Glycol; 0.0% Xylene

Formula: C₂₀H₁₉ClN₄
Density: 1 g/cm³ (8.345 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR