Ethylene glycol diglycidyl ether

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Quetol 651, 1,2-Diglycidyloxyethane, 1,2-Bis(2,3-epoxypropoxy)ethane

20440
100504-256EA 349.2 USD
100504-256
Ethylene glycol diglycidyl ether
Ethylene glycol diglycidyl ether

Quetol 651, Resin

Formula: C₈H₁₄O₄
MW: 174.19 g/mol
Boiling Pt: 112 °C (234 °F)
Flash Pt: 110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005142
CAS Number: 2224-15-9
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5b0730808ffe7c537a32d6c48a88c78b052e042&vid=d5b0730808ffe7c537a32d6c48a88c78b052e042&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Specific gravity (H₂O=1) 1.15

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR