L(+)-Potassium sodium tartrate tetrahydrate 99.0-102.0% ACS

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Potassium sodium tartrate tetrahydrate, Potassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate, Potassium sodium L(+)-tartrate tetrahydrate, Sodium potassium tartrate, (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrate, (R,R)-(+)-Potassium sodium tartrate tetrahydrate, Rochelle salt, L(+)-Tartaric acid potassium sodium salt tetrahydrate, (+)-Potassium sodium L-tartrate tetrahydrate

20260
100504-246EA 80.65 USD
100504-246
L(+)-Potassium sodium tartrate tetrahydrate 99.0-102.0% ACS
L(+)-Potassium sodium tartrate tetrahydrate

Used with bismuth subnitrate for staining polysaccharides, including glycogen without oxidizing agents.


Reference: Shinji, Y., et al (1975). A new electron microscopic histo-cyto chemical staining method: demonstration of glycogen particles. Acta Histochem., Cytochem., 8:139.

Formula: KOOC(CHOH)₂COONa·4H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150989
CAS Number: 6381-59-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d585522f4e3b14a7b011dc8a858f5eeca413864&vid=542d22225b124dda886e5007091b35db67214bb7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0-102.0%
Water 21.0-27.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR