Potassium hexacyanoferrate(III) ACS

Supplier: Electron Microscopy Sciences
Warning

20150
100504-230EA 101.83 USD
100504-230
Potassium hexacyanoferrate(III) ACS
Potassium hexacyanoferrate (III)
Formula: K₃Fe(CN)₆
Density: 1.89 g/cm³ (15.772 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2

Order Now

Specification Test Results

Chloride 0.01%
Ferro compounds (Fe(Cn)₄ 0.05%
Insoluble matters 0.005%
Sulfate 0.01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR