Potassium dichromate, crystals ACS

Supplier: Electron Microscopy Sciences
Danger

20100
100504-228EA 83.77 USD
100504-228
Potassium dichromate, crystals ACS
Potassium dichromate
Formula: K₂Cr₂O₇
Melting Pt: 398 °C (748 °F)
Density: 2.68 g/cm³ (22.365 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
UN: 3086
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee9e04c3058dfd245fb781403d336f0fc2c3492f&vid=ee9e04c3058dfd245fb781403d336f0fc2c3492f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Insoluble and NH₄OH ppt. 0.005%
Chloride 0.001%
Sulfate 0.005%
Calcium 0.003%
Sodium 0.02%
Loss on drying (105 °C) 0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR