Potassium chloride, crystals ACS

Supplier: Electron Microscopy Sciences
Warning

20060
100504-226EA 31.75 USD
100504-226
Potassium chloride, crystals ACS
Potassium chloride

KCl F.W. 74.55

Formula: KCl
Boiling Pt: 1500 °C (2732 °F)
Melting Pt: 770 °C (1418 °F)
Density: 1.98 g/cm³ (16.523 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07dabac366fc2e211f34c0a1eff3242514e3b0cd&vid=07dabac366fc2e211f34c0a1eff3242514e3b0cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Bromide 0.01%
Chlorate and nitrate 0.003%
Insoluble matters 0.005%
Iodide 0.002%
Phosphate 0.0005%
Sodium 0.005%
Sulfate 0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR