Polyethylene glycol 1,000 waxy

Supplier: Electron Microscopy Sciences

Synonyms: PEG 1,000, PEG 900-1100

19740
100504-212EA 116.97 USD
100504-212
Polyethylene glycol 1,000 waxy
Polyethylene glycol 1,000

An embedding medium for microscopy and histochemistry

Formula: H(OCH₂CH₂)nOH
Density: 1.0926 (60 °C)
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR