Polyethylene glycol 400 viscous liquid

Supplier: Electron Microscopy Sciences

Synonyms: PEG 400

19720
100504-208EA 60.97 USD
100504-208
Polyethylene glycol 400 viscous liquid
Polyethylene glycol 400

An embedding medium for microscopy and histochemistry

Formula: H(OCH₂CH₂)nOH
Density: 1.1254 (20 °C)
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR